![[Val5]-Angiotensin II acetate salt hydrate ≥95% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/301/478/c093dfb8-a891-46ac-b6a1-655b77da7c03/640/c093dfb8-a891-46ac-b6a1-655b77da7c03.png)
[Val5]-Angiotensin II acetate salt hydrate
![[Val5]-Angiotensin II acetate salt hydrate ≥95% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/301/478/c093dfb8-a891-46ac-b6a1-655b77da7c03/640/c093dfb8-a891-46ac-b6a1-655b77da7c03.png)
Empirical Formula (Hill Notation):
C49H69N13O12 · xC2H4O2 · yH2O
Molecular Weight:
1032.15 (anhydrous free base basis)
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing