
Triamterene Related Compound A

Synonym(s):
2,4,6-Triamino-5-nitrosopyrimidine
Empirical Formula (Hill Notation):
C4H6N6O
CAS Number:
Molecular Weight:
154.13
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2