
Plastic additive 26

Synonym(s):
Trioctyl trimellitate, Tris(2-ethylhexyl) trimellitate
Linear Formula:
C6H3-1,2,4-[CO2CH2CH(C2H5)(CH2)3CH3]3
CAS Number:
Molecular Weight:
546.78
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing