![Phosphocholine chloride calcium salt tetrahydrate Sigma Grade](/deepweb/assets/sigmaaldrich/product/structures/286/996/ef5a69df-52a3-49ae-b828-b53dbe52f555/640/ef5a69df-52a3-49ae-b828-b53dbe52f555.png)
Phosphocholine chloride calcium salt tetrahydrate
![Phosphocholine chloride calcium salt tetrahydrate Sigma Grade](/deepweb/assets/sigmaaldrich/product/structures/286/996/ef5a69df-52a3-49ae-b828-b53dbe52f555/640/ef5a69df-52a3-49ae-b828-b53dbe52f555.png)
Synonym(s):
Calcium phosphorylcholine chloride, Choline chloride phosphate calcium salt, Choline phosphate chloride, Phosphorocholine chloride calcium salt, Phosphorylcholine chloride calcium salt
Linear Formula:
CaO3POCH2CH2N(Cl)(CH3)3 · 4H2O
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing