
Oseltamivir impurity H

Synonym(s):
Tributylphosphine oxide, Butyphos
Linear Formula:
[CH3(CH2)3]3P(O)
CAS Number:
Molecular Weight:
218.32
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing