
Minocycline for system suitability

Synonym(s):
Minocycline hydrochloride
Empirical Formula (Hill Notation):
C23H27N3O7 · HCl
CAS Number:
Molecular Weight:
493.94
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing