
L-Proline-13C5

Empirical Formula (Hill Notation):
13C5H9NO2
CAS Number:
Molecular Weight:
120.09
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing