
Lithium titanate

Synonym(s):
LTO, Lithium titanate spinel oxide
Linear Formula:
Li4Ti5O12
Products
Product Category
Brand
Physical Form
Chemical Composition
Quality Segment
Application
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing