
Geniposide

Synonym(s):
(1S,4αS,7αS)- 1-(β-D-Glucopyranosyloxy)-1,4a,5,7a-tetrahydro-7-(hydroxymethyl)- cyclopenta[c]pyran-4-carboxylic acid methyl ester
Empirical Formula (Hill Notation):
C17H24O10
CAS Number:
Molecular Weight:
388.37
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing