
Disodium Inosinate

Synonym(s):
Inosine 5′-monophosphate disodium salt hydrate, 5′-IMP-Na2, 5′-Inosinic acid disodium salt hydrate, I-5′-P, IMP, Inosinic Acid
Empirical Formula (Hill Notation):
C10H11N4O8PNa2 · xH2O
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2