
Digoxigenin

Synonym(s):
3β,12β,14β,21-Tetrahydroxy-20(22)-norcholenic acid lactone, 3β,12β,14-Trihydroxy-5β,20(22)-cardenolide, 5β,20(22)-Cardenolide-3β,12β,14-triol, Lanadigigenin
Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Functional Group
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing