Skip to Content
Merck
vibrant-m

Clomipramine Related Compound D

vibrant-m
Synonym(s):
3-(3,7-Dichloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine hydrochloride
Empirical Formula (Hill Notation):
C19H22Cl2N2 · HCl
CAS Number:
Molecular Weight:
385.76

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1