
Calcineurin from bovine brain

Synonym(s):
Calcium/Calmodulin-Activated Protein Phosphatase, Calmodulin binding protein, Modulator binding protein, PP2B, Phosphoprotein Phosphohydrolase, Protein phosphatase 2B
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing