
Bufalin

Synonym(s):
3β,14-Dihydroxy-5β,20(22)-bufadienolide, 5β,20(22)-Bufadienolide-3β,14-diol, Bufalin
Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3