Skip to Content
Merck
Boc-Orn(Z)-OH ≥98.0% (TLC)

Boc-Orn(Z)-OH

Boc-Orn(Z)-OH ≥98.0% (TLC)
Synonym(s):
Nα-Boc-Nδ-Cbz-L-ornithine, Nα-Boc-Nδ-Z-L-ornithine, Nδ-Z-Nα-Boc-L-ornithine, Boc-Orn(Z)-OH
Empirical Formula (Hill Notation):
C18H26N2O6
CAS Number:
Molecular Weight:
366.41

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing