
Anti-Human Polyvalent Immunoglobulins (G,A,M) antibody produced in goat

Products
Product Category
Brand
Biological Source
Clonality
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing