Skip to Content
Merck
vibrant-m

1,4-Dichlorobenzene solution

vibrant-m
Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00

Products

Product Category
Brand
Formula Weight
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1