10-Methoxycarbamazepine
Synonym(s):
10-Methoxy-5H-dibenz[b,f]azepine-5-carboxamide
Empirical Formula (Hill Notation):
C16H14N2O2
CAS Number:
Molecular Weight:
266.29
Products
Brand
Formula Weight
Quality Segment
Application
Markush Class
Markush Group
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing