Skip to Content
Merck
All Photos(1)

Key Documents

51016

Sigma-Aldrich

Atto 550 DPPE

suitable for fluorescence

Synonym(s):

1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine labeled with Atto 550

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

Assay

≥90% (TLC)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

λ

in chloroform/methanol (8/2)

UV absorption

λ: 559-565 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 550 is a fluorescent label related to the well-known dye Rhodamine 6G. The dye is designed for application in the area of life science, e.g. labeling of DNA, RNA or proteins. Characteristic features of the label are strong absorption, high fluorescence quantum yield, high thermal and photo-stability, and very little triplet formation. The dye is highly suitable for single-molecule detection applications and high-resolution microscopy.
Atto 550 is a cationic dye (charge +1). As supplied Atto 550 consists of three isomers with identical absorption and fluorescence.

Atto-Dye Labeled Phospholipids
Sigma-Aldrich offers a variety of glycero-phospholipids carrying one or two fatty acid groups (lipophilic groups) and a phosphate ester residue (hydrophilic group). They are labeled at the hydrophilic head group. After incorporation of the phospholipid into a membrane the fluorophore is located at the water/lipid interface of the membrane. We currently provide 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine (DPPE), 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE), palmitoyl-sn-glycero-phosphoethanolamine (PPE), and 1,2-dimyristoyl-sn-glycero-3-phospho-ethanolamine (DMPE) labeled with Atto-dyes.
find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service