Skip to Content
Merck
All Photos(2)

Key Documents

07-672-I

Sigma-Aldrich

Anti-MYPT1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein phosphatase 1 regulatory subunit 12A, 130 kDa myosin-binding subunit of smooth muscle myosin phosphatase, Myosin phosphatase-targeting subunit 1, Myosin phosphatase target subunit 1, PP1M subunit M110, Protein phosphatase myosin-binding subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chicken (immunogen homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPP1R12A(4659)

General description

Protein phosphatase 1 regulatory subunit 12A (UniProt: Q90623; also known as 130 kDa myosin-binding subunit of smooth muscle myosin phosphatase, Myosin phosphatase-targeting subunit 1, Myosin phosphatase target subunit 1, PP1M subunit M110, Protein phosphatase myosin-binding subunit, MYPT1) is encoded by the PPP1R12A (also known as MBS, MYPT1) gene (Gene ID: 396020) in human. MYPT1 is one of the subunits and an integral component of the Myosin phosphatase. It is detected in brain, lung, aorta, heart, gizzard, stomach, oviduct, spleen, kidney, and small intestine. MYPT1 is localized on stress fibers, and is distributed close to the cell membrane and at cell-cell contacts to regulate Myosin phosphatase activity. It is phosphorylated by Rho-associated kinases on serine and threonine residues. Phosphorylation at Threonine 695 is shown to inhibit myosin phosphatase activity and phosphorylation at Threonine 850 is reported to abolish myosin binding. Two isoforms of MYPT1 have been reported that are produced by alternative splicing.

Specificity

This rabbit polyclonal antibody detects human Protein phosphatase 1 regulatory subunit 12A. It targets an epitope within 291 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of chicken MYPT1.

Application

Detect MYPT1 using this rabbit polyclonal antibody, Anti-MYPT1 Antibody validated for use in western blotting & IP.
Immunoprecipitation Analysis: 0.5µg from a representative lot immunoprecipitated MYPT1 from 0.5mg of HeLa cell lysate.
Research Category
Signaling
Research Sub Category
Metabolic Hormones & Receptors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MYPT1 in 10 µg of HeLa cell lysate.

Target description

~120/130 kDa observed; 111.61 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 04-386

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Upendarrao Golla et al.
Cancers, 13(19) (2021-10-14)
The poor prognosis of acute myeloid leukemia (AML) and the highly heterogenous nature of the disease motivates targeted gene therapeutic investigations. Rho-associated protein kinases (ROCKs) are crucial for various actin cytoskeletal changes, which have established malignant consequences in various cancers
Sara Basbous et al.
Cell death & disease, 15(1), 46-46 (2024-01-14)
Entosis is a process that leads to the formation of cell-in-cell structures commonly found in cancers. Here, we identified entosis in hepatocellular carcinoma and the loss of Rnd3 (also known as RhoE) as an efficient inducer of this mechanism. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service