Merck
Inhibition of GABA transporters fails to afford significant protection following focal cerebral ischemia.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2017-11-18)
Maria Ek Lie, Emma K Gowing, Rasmus P Clausen, Petrine Wellendorph, Andrew N Clarkson
RÉSUMÉ
Brain ischemia triggers excitotoxicity and cell death, yet no neuroprotective drugs have made it to the clinic. While enhancing GABAergic signaling to counterbalance excitotoxicity has shown promise in animal models, clinical studies have failed. Blockade of GABA transporters (GATs) offers an indirect approach to increase GABA inhibition to lower the excitation threshold of neurons. Among the GATs, GAT1 is known to promote neuroprotection, while the protective role of the extrasynaptic transporters GAT3 and BGT1 is elusive. A focal lesion was induced in the motor cortex in two to four-month-old C57BL/6 J male mice by photothrombosis. The GAT1 inhibitor, tiagabine (1 and 10 mg/kg), the GAT2/3 inhibitor, ( S)-SNAP-5114 (5 and 30 mg/kg) and the GAT1/BGT1 inhibitor, EF-1502 (1 and 10 mg/kg) were given i.p. 1 and 6 h post-stroke to assess their impact on infarct volume and motor performance seven days post-stroke. One mg/kg tiagabine improved motor performance, while 10 mg/kg tiagabine, ( S)-SNAP-5114 and EF-1502 had no effect. None of the compounds affected infarct volume. Interestingly, treatment with tiagabine induced seizures and ( S)-SNAP-5114 led to increased mortality. Although we show that tiagabine can promote protection, our findings indicate that caution should be had when using GAT1 and GAT3 inhibitors for conditions of brain ischemia.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(S)-SNAP-5114, ≥98% (HPLC), solid