Merck
Systemic candesartan reduces brain angiotensin II via downregulation of brain renin-angiotensin system.
Hypertension research : official journal of the Japanese Society of Hypertension (2009-11-28)
Nicolas Pelisch, Naohisa Hosomi, Masaki Ueno, Hisashi Masugata, Koji Murao, Hirofumi Hitomi, Daisuke Nakano, Hiroyuki Kobori, Akira Nishiyama, Masakazu Kohno
RÉSUMÉ
The renin-angiotensin system has an important function in the regulation of blood pressure as well as in pathophysiological processes in the central nervous system. We examined the effects of the angiotensin receptor blocker candesartan (10 mg kg(-1) per day, p.o.) on brain angiotensin II levels in angiotensin II-infused hypertensive rats. Angiotensin II or vehicle was infused subcutaneously for 14 days in Sprague-Dawley rats. Angiotensin II infusion resulted in increased blood pressure, an effect that was blocked by candesartan treatment. There was no effect of the angiotensin II infusion on Angiotensin II levels in the brain or on blood-brain barrier permeability. Brain tissue angiotensinogen and angiotensin converting enzyme mRNA levels were not changed by angiotensin II infusion but were decreased by candesartan treatment. At 2 weeks of treatment, CV11974, an active form of candesartan, was detectable in the plasma but was not detectable in brain tissue. These data suggest that treatment with candesartan decreases brain angiotensin II by decreasing brain angiotensinogen and angiotensin converting enzyme gene expression.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Angiotensine II humaine, ≥93% (HPLC), powder
Sigma-Aldrich
[Val5]-Angiotensin II acetate salt hydrate, ≥95% (HPLC), powder