Merck
  • Presence of sensory nerve corpuscles in the human corpus and cervix uteri during pregnancy and labor as revealed by immunohistochemistry.
Presence of sensory nerve corpuscles in the human corpus and cervix uteri during pregnancy and labor as revealed by immunohistochemistry.
Reproductive biology and endocrinology : RB&E (2006-08-30)
Berith K Tingaker, Gunvor Ekman-Ordeberg, Sture Forsgren
RÉSUMÉ
The uterus is exposed to changes such as enlargement and distension during pregnancy and labor. In these processes and in the process of cervical ripening, proprioceptive information is likely to be of great importance. Therefore, we wanted to study the possible existence of sensory nerve corpuscles in uterine corpus and cervix during pregnancy and labor. Studies on this aspect have not previously been perfomed. Biopsies were taken from the upper edge of the hysterotomy during caesarean section at term (n = 8), in labor (n = 5) and from the corresponding area in the non-pregnant uterus after hysterectomy (n = 7). Cervical biopsies were obtained transvaginally from the anterior cervical lip. Serial cryostat sections were prepared for immunohistochemistry using polyclonal antibodies against nerve growth factor receptor p75, protein gene product 9.5 and S-100. Structures with the characteristics of sensory nerve corpuscles were observed in several specimens after staining for p75, PGP 9.5 and S-100. They were observed in specimens of the non-pregnant corpus and cervix and also in specimens of the pregnant cervix before onset of labor. However, they were absent in all specimens during labor. Sensory corpuscles have here for the first time been detected in the human corpus and cervix uteri. Studies on the importance of the corpuscles in relation to the protective reflex actions that occur in the uterus during pregnancy should be performed in the future.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-S-100 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Anti-Nerve Growth Factor Receptor (NGFR p75) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Référence
Conditionnement
Disponibilité
Prix
Quantité