Merck
Bromoisobutyramide as an intermolecular surface binder for the preparation of free-standing biopolymer assemblies.
Advanced materials (Deerfield Beach, Fla.) (2011-11-22)
Damien Mertz, Pramudana Tan, Yajun Wang, Tor Kit Goh, Anton Blencowe, Frank Caruso
RÉSUMÉ
Bromoisobutyramide (BrIBAM)-modified silica templates facilitate the formation of bio-functional thin films made of a range of biopolymers (e.g., polypeptides, nucleic acids or polysaccharides). Upon template removal, non-covalent free-standing biopolymeric assemblies (e.g., hollow capsules or replicated spheres and fibers) are formed without the need for covalent cross-linking.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Isobutyramide, 99%