Merck
Polycomb deficiency drives a FOXP2-high aggressive state targetable by epigenetic inhibitors.
Nature communications (2023-01-21)
Fan Chen, Aria L Byrd, Jinpeng Liu, Robert M Flight, Tanner J DuCote, Kassandra J Naughton, Xiulong Song, Abigail R Edgin, Alexsandr Lukyanchuk, Danielle T Dixon, Christian M Gosser, Dave-Preston Esoe, Rani D Jayswal, Stuart H Orkin, Hunter N B Moseley, Chi Wang, Christine Fillmore Brainson
RÉSUMÉ
Inhibitors of the Polycomb Repressive Complex 2 (PRC2) histone methyltransferase EZH2 are approved for certain cancers, but realizing their wider utility relies upon understanding PRC2 biology in each cancer system. Using a genetic model to delete Ezh2 in KRAS-driven lung adenocarcinomas, we observed that Ezh2 haplo-insufficient tumors were less lethal and lower grade than Ezh2 fully-insufficient tumors, which were poorly differentiated and metastatic. Using three-dimensional cultures and in vivo experiments, we determined that EZH2-deficient tumors were vulnerable to H3K27 demethylase or BET inhibitors. PRC2 loss/inhibition led to de-repression of FOXP2, a transcription factor that promotes migration and stemness, and FOXP2 could be suppressed by BET inhibition. Poorly differentiated human lung cancers were enriched for an H3K27me3-low state, representing a subtype that may benefit from BET inhibition as a single therapy or combined with additional EZH2 inhibition. These data highlight diverse roles of PRC2 in KRAS-driven lung adenocarcinomas, and demonstrate the utility of three-dimensional cultures for exploring epigenetic drug sensitivities for cancer.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Toxine cholérique from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Anticorps anti-triméthyl-histone H3 (Lys27), Upstate®, from rabbit
Sigma-Aldrich
Anti-FOXP2 antibody produced in rabbit, AB1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution