Merck
Metabolic targeting synergizes with MAPK inhibition and delays drug resistance in melanoma.
Cancer letters (2018-11-28)
Christina Brummer, Stephanie Faerber, Christina Bruss, Christian Blank, Ruben Lacroix, Sebastian Haferkamp, Wolfgang Herr, Marina Kreutz, Kathrin Renner
RÉSUMÉ
Tumors, including melanomas, frequently show an accelerated glucose metabolism. Mutations in the v-Raf murine sarcoma viral oncogene homolog B (BRAF), detected in about 50% of all melanomas, result in further enhancement of glycolysis. Therefore anti-metabolic substances might enhance the impact of RAF inhibitors. We have identified the two non-steroidal anti-inflammatory drugs (NSAIDs) diclofenac and lumiracoxib being able to restrict energy metabolism in human melanoma cells by targeting lactate release and oxidative phosphorylation (OXPHOS). In combination with the RAF inhibitor vemurafenib strong synergism was observed: Diclofenac as well as lumiracoxib increased the anti-glycolytic impact of vemurafenib and prevented RAF-inhibitor induced metabolic reprogramming towards OXPHOS. Consequently, both NSAIDs sensitized melanoma cells to vemurafenib triggered proliferation arrest and enhanced the anti-tumor effect of RAF inhibitors from cytostatic to cytotoxic. Furthermore the addition of NSAIDs delayed the onset of RAF inhibitor resistance, most likely by counteracting the upregulation of MITF. Our data suggest that selected NSAIDs could be a promising combination partner for MAPK pathway inhibitors for the treatment of BRAFV600E mutated melanomas.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Lumiracoxib, ≥98% (HPLC)