- The Difference in Cytotoxic Activity between Two Optical Isomers of Gelsemine from Gelsemium elegans Benth. on PC12 Cells.
The Difference in Cytotoxic Activity between Two Optical Isomers of Gelsemine from Gelsemium elegans Benth. on PC12 Cells.
Molecules (Basel, Switzerland) (2019-05-28)
Li Lin, Yan-Chun Liu, Zhao-Ying Liu
PMID31130596
RÉSUMÉ
Two optical isomers, +/- gelsemine (1, 2), together with one known compound were isolated from the whole plant of G. elegans. The structures of the separated constituents were elucidated on 1D and 2D (1H-1H COSY, HMBC, HSQC) NMR spectroscopy and high-resolution mass spectrometry (HRMS). The isolated alkaloids were tested in vitro for cytotoxic potential against PC12 cells by the MTT assay. As a result, (+) gelsemine (compound 1) exhibited cytotoxic activity against PC12 cells with an IC50 value of 31.59 μM, while (-) gelsemine (compound 2) was not cytotoxic.