Merck
Huntingtin promotes cell survival by preventing Pak2 cleavage.
Journal of cell science (2009-02-26)
Shouqing Luo, David C Rubinsztein
RÉSUMÉ
Huntington's disease is caused by a polyglutamine expansion in the huntingtin protein. Wild-type huntingtin, by contrast, appears to protect cells from pro-apoptotic insults. Here we describe a novel anti-apoptotic function for huntingtin. When cells are exposed to Fas-related signals, the ubiquitously expressed p21-activated kinase 2 (Pak2) can be activated via cleavage by caspases to release a constitutively active C-terminal fragment, which mediates cell death. Our data show that huntingtin interacts with Pak2. Overexpression of huntingtin significantly inhibits caspase-3-mediated and caspase-8-mediated cleavage of Pak2 in cells. Moreover, huntingtin prevents Pak2 cleavage by caspase-3 and caspase-8 in vitro. Although huntingtin is cytoprotective in wild-type cells that are exposed to TNFalpha, it has no significant benefit in TNFalpha-treated cells with Pak2 knockdown. Thus, huntingtin exerts anti-apoptotic effects by binding to Pak2, which reduces the abilities of caspase-3 and caspase-8 to cleave Pak2 and convert it into a mediator of cell death.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-protéine huntingtine, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Caspase-3 Cellular Activity Assay Kit
Référence
Conditionnement
Disponibilité
Prix
Quantité