Principaux documents
Z552410
Sigma-Aldrich
Aldrich® high-efficiency Friedrichs reflux condenser
joint: ST/NS 24/40
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
Description
glass hose barbs
Longueur
220 mm
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC