Principaux documents
DWK11-0053B
Wheaton® Microliter Snap Caps With Septa, Light Blue Cap, Gc Only
light blue PTFE/silicone cap, GC only
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.11
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
light blue PTFE/silicone cap
Conditionnement
pkg of 100 ea
Fabricant/nom de marque
WHEATON® 11-0053B
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 4
Cet article
DWK11-0055BDWK11-0051GDWK11-0051K
manufacturer/tradename
WHEATON® 11-0053B
manufacturer/tradename
WHEATON® 11-0055B
manufacturer/tradename
WHEATON® 11-0051G
manufacturer/tradename
WHEATON® 11-0051K
packaging
pkg of 100 ea
packaging
pkg of 100 ea
packaging
pkg of 100 ea
packaging
pkg of 100 ea
Application
Chromatography
Caractéristiques et avantages
<ul><li>11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.</li><li>Where noted, the SnapTin Ring is installed between cap and septa for robotics employing magnetics to move vials.</li><li>Approved for GC and LC (where noted).</li></ul>
  • 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
  • Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
  • Approved for GC and LC (where noted).
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.
  • Faites votre choix parmi les versions les plus récentes :