Principaux documents
BAF196030000
SP Bel-Art® “T” Shaped Tubing Connectors
for ³⁄₁₆ in. Tubing
Synonyme(s) :
connectors, tubing, tubing coupler
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Bel-Art F19603-0000
D.I.
4.7 mm (0.1875 in.)
L × largeur
1.5 in. (38.1 mm) × 0.875 in. (22.2 mm)
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 4
Cet article
BAF196040000BAF196060000BAF196130000
packaging
pack of 12 ea
packaging
pack of 12 ea
packaging
pack of 12 ea
packaging
pack of 12 ea
I.D.
4.7 mm (0.1875 in.)
I.D.
-
I.D.
-
I.D.
-
L × width
1.5 in. (38.1 mm) × 0.875 in. (22.2 mm)
L × width
-
L × width
-
L × width
-
Description générale
Connectors have multiple barbs per arm providing additional contact between the fitting and the tubing for extra gripping power.
Caractéristiques et avantages
  • Ensures a tight, secure connection between lengths of tubing having the same inner diameter (I.D.)
  • Chemical-resistant polypropylene for use with a wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP