HPLC Analysis of Omeprazole Enantiomers on Astec® CYCLOBOND I 2000 DMP
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
[A] acetonitrile; [B] acetic acid; [C] ammonium hydroxide; (100:0.4:0.1, A:B:C)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 302 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null