HPLC Analysis of Norphenylephrine Enantiomers on Astec® CYCLOBOND I 2000 AC (pH 4.1)
CONDITIONS
column
CYCLOBOND I 2000 AC, 25 cm x 4.6 mm I.D., 5 μm particles (20124AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (93:7, A:B)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null