Merck
  • GPR55 agonist lysophosphatidylinositol and lysophosphatidylcholine inhibit endothelial cell hyperpolarization via GPR-independent suppression of Na
GPR55 agonist lysophosphatidylinositol and lysophosphatidylcholine inhibit endothelial cell hyperpolarization via GPR-independent suppression of Na
Vascular pharmacology (2017-01-09)
Alexander I Bondarenko, Fabrizio Montecucco, Olga Panasiuk, Vadim Sagach, Nataliya Sidoryak, Karim J Brandt, François Mach
ABSTRACT
Lysophosphatidylinositol (LPI) and lysophosphatidylcholine (LPC) are lipid signaling molecules that induce endothelium-dependent vasodilation. In addition, LPC suppresses acetylcholine (Ach)-induced responses. We aimed to determine the influence of LPC and LPI on hyperpolarizing responses in vitro and in situ endothelial cells (EC) and identify the underlying mechanisms. Using patch-clamp method, we show that LPI and LPC inhibit EC hyperpolarization to histamine and suppress Na
Sigma-Aldrich
SKA-31, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
400 μL
Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$496.00