Merck
  • Purification and properties of extracellular signal-regulated kinase 1, an insulin-stimulated microtubule-associated protein 2 kinase.
Purification and properties of extracellular signal-regulated kinase 1, an insulin-stimulated microtubule-associated protein 2 kinase.
Biochemistry (1991-01-08)
T G Boulton, J S Gregory, M H Cobb
ABSTRACT
In rat 1 fibroblasts, insulin has little or no stimulatory effect on the activities of either MAP2 protein kinase or ribosomal protein S6 kinase. In contrast, in rat 1 cells that overexpress the normal human insulin receptor (rat 1 HIRc B; McClain et al. (1987) J. Biol. Chem. 262, 14663-14671), insulin activates both MAP2 and S6 kinase activities close to 5-fold. A MAP2 kinase has been purified from insulin-treated rat 1 HIRc B cells over 6300-fold by chromatography on Q-Sepharose, phenyl-Sepharose, S-Sepharose, phosphocellulose, QAE-Sepharose, UltrogelAcA54, DEAE-cellulose, and a second Q-Sepharose. Its specific activity is approximately 0.8-1 mumol.min-1.mg-1 with MAP2 and 3 mumol.min-1.mg-1 with myelin basic protein. The enzyme preparation contains one major band of Mr = 43,000 upon SDS-polyacrylamide gel electrophoresis, which is immunoblotted by antibodies to phosphotyrosine. A sequence from the 43-kDa band led to the isolation of a cDNA encoding the enzyme, which we have named ERK1 for extracellular signal-regulated kinase (Boulton et al. (1990) Science 249, 64-67).
Sigma-Aldrich
ERK1, active, untagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
ERK2, active, GST tagged human, recombinant, expressed in E. coli, PRECISIO® Kinase, ≥70% (SDS-PAGE), buffered aqueous glycerol solution