Merck
  • Identification of p62/SQSTM1 as a component of non-canonical Wnt VANGL2-JNK signalling in breast cancer.
Identification of p62/SQSTM1 as a component of non-canonical Wnt VANGL2-JNK signalling in breast cancer.
Nature communications (2016-01-13)
Tania M Puvirajesinghe, François Bertucci, Ashish Jain, Pierluigi Scerbo, Edwige Belotti, Stéphane Audebert, Michael Sebbagh, Marc Lopez, Andreas Brech, Pascal Finetti, Emmanuelle Charafe-Jauffret, Max Chaffanet, Rémy Castellano, Audrey Restouin, Sylvie Marchetto, Yves Collette, Anthony Gonçalvès, Ian Macara, Daniel Birnbaum, Laurent Kodjabachian, Terje Johansen, Jean-Paul Borg
ABSTRACT
The non-canonical Wnt/planar cell polarity (Wnt/PCP) pathway plays a crucial role in embryonic development. Recent work has linked defects of this pathway to breast cancer aggressiveness and proposed Wnt/PCP signalling as a therapeutic target. Here we show that the archetypal Wnt/PCP protein VANGL2 is overexpressed in basal breast cancers, associated with poor prognosis and implicated in tumour growth. We identify the scaffold p62/SQSTM1 protein as a novel VANGL2-binding partner and show its key role in an evolutionarily conserved VANGL2-p62/SQSTM1-JNK pathway. This proliferative signalling cascade is upregulated in breast cancer patients with shorter survival and can be inactivated in patient-derived xenograft cells by inhibition of the JNK pathway or by disruption of the VANGL2-p62/SQSTM1 interaction. VANGL2-JNK signalling is thus a potential target for breast cancer therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human SQSTM1
Sigma-Aldrich
MISSION® esiRNA, targeting human VANGL2