Merck
  • Interference of flavonoids with enzymatic assays for the determination of free fatty acid and triglyceride levels.
Interference of flavonoids with enzymatic assays for the determination of free fatty acid and triglyceride levels.
Analytical and bioanalytical chemistry (2011-11-29)
Elise F Hoek-van den Hil, Karsten Beekmann, Jaap Keijer, Peter C H Hollman, Ivonne M C M Rietjens, Evert M van Schothorst
ABSTRACT
Flavonoids are bioactive food compounds with potential lipid-lowering effects. Commercially available enzymatic assays are widely used to determine free fatty acid (FFA) and triglyceride (TG) levels both in vivo in plasma or serum and in vitro in cell culture medium or cell lysate. However, we have observed that various flavonoids interfere with peroxidases used in these enzymatic assays, resulting in incorrect lower FFA and TG levels than actually present. Furthermore, addition of isorhamnetin or the major metabolite of the flavonoid quercetin in human and rat plasma, quercetin-3-O-glucuronide, to murine serum also resulted in a significant reduction of the detected TG levels, while a trend was seen for FFA levels. It is concluded that when applying these assays, vigilance is needed and alternative analytical methods, directly assessing FFA or TG levels, should be used for studying the biological effects of flavonoids on FFA and TG levels.
Product Description
Supelco
Isorhamnetin, analytical standard
Sigma-Aldrich
Isorhamnetin, ≥95.0% (HPLC)