Merck
Attending rounds: patient with hypokalemia and metabolic acidosis.
Clinical journal of the American Society of Nephrology : CJASN (2011-09-17)
Asghar Rastegar
ABSTRACT
Hypokalemic paralysis represents a medical emergency requiring both rapid diagnosis and treatment. In this Attending Rounds a patient with hypokalemia and metabolic acidosis is presented to emphasize the role of routine laboratory studies in the assessment of such patients so that a correct diagnosis can be made and appropriate treatment can be initiated promptly.
Sigma-Aldrich
Potassium citrate tribasic monohydrate, ≥98% (GC/titration)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, suitable for cell culture
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets analytical specification of Ph. Eur., BP, FCC, E332, 99-100.5% (calc with ref. to anhyd. subst.)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets USP testing specifications