Merck
  • Antihyperlipidemic effect of D-pinitol on streptozotocin-induced diabetic Wistar rats.
Antihyperlipidemic effect of D-pinitol on streptozotocin-induced diabetic Wistar rats.
Journal of biochemical and molecular toxicology (2008-08-30)
P K M Anu Geethan, P Stanely Mainzen Prince
ABSTRACT
D-pinitol (3-O-methyl-chiroinositol), an active principle of the traditional antidiabetic plant, Bougainvillea spectabilis, is claimed to exert insulin-like effects. This study was undertaken to evaluate the effect of D-pinitol on lipids and lipoproteins in streptozotocin (STZ)-induced diabetic Wistar rats. Rats were made type II diabetic by single intraperitoneal injection of STZ at a dose of 40 mg/kg body weight. STZ-induced diabetic rats showed significant (p < 0.05) increase in the levels of blood glucose and total cholesterol, triglycerides, free fatty acids, and phospholipids in serum, liver, kidney, heart, and brain. The levels of low-density lipoprotein (LDL) and very low-density lipoprotein (VLDL) cholesterol were significantly increased, and the level of high-density lipoprotein (HDL) cholesterol was significantly decreased in diabetic rats Oral administration of D-pinitol to STZ-induced diabetic rats showed significant (p < 0.05) decrease in the levels of blood glucose and total cholesterol, triglycerides, free fatty acids, and phospholipids in serum, liver, kidney, heart, and brain. The D-pinitol also lowered significantly (p < 0.05) LDL and VLDL cholesterol levels and increased significantly (p < 0.05) HDL cholesterol levels in the serum of diabetic rats. Thus, the present study clearly showed the antihyperlipidemic effect of D-pinitol in STZ-induced type II diabetic rats.
Product Description
Supelco
D-Pinitol, analytical standard
Sigma-Aldrich
D-Pinitol, 95%