Merck
  • A new family of covalent inhibitors block nucleotide binding to the active site of pyruvate kinase.
A new family of covalent inhibitors block nucleotide binding to the active site of pyruvate kinase.
The Biochemical journal (2012-08-22)
Hugh P Morgan, Martin J Walsh, Elizabeth A Blackburn, Martin A Wear, Matthew B Boxer, Min Shen, Henrike Veith, Iain W McNae, Matthew W Nowicki, Paul A M Michels, Douglas S Auld, Linda A Fothergill-Gilmore, Malcolm D Walkinshaw
ABSTRACT
PYK (pyruvate kinase) plays a central role in the metabolism of many organisms and cell types, but the elucidation of the details of its function in a systems biology context has been hampered by the lack of specific high-affinity small-molecule inhibitors. High-throughput screening has been used to identify a family of saccharin derivatives which inhibit LmPYK (Leishmania mexicana PYK) activity in a time- (and dose-) dependent manner, a characteristic of irreversible inhibition. The crystal structure of DBS {4-[(1,1-dioxo-1,2-benzothiazol-3-yl)sulfanyl]benzoic acid} complexed with LmPYK shows that the saccharin moiety reacts with an active-site lysine residue (Lys335), forming a covalent bond and sterically hindering the binding of ADP/ATP. Mutation of the lysine residue to an arginine residue eliminated the effect of the inhibitor molecule, providing confirmation of the proposed inhibitor mechanism. This lysine residue is conserved in the active sites of the four human PYK isoenzymes, which were also found to be irreversibly inhibited by DBS. X-ray structures of PYK isoforms show structural differences at the DBS-binding pocket, and this covalent inhibitor of PYK provides a chemical scaffold for the design of new families of potentially isoform-specific irreversible inhibitors.
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Suramin sodium salt, ≥98% (TLC)
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)