Merck
  • Human Minor Salivary Glands: A Readily Available Source of Salivary Stem/Progenitor Cells for Regenerative Applications.
Human Minor Salivary Glands: A Readily Available Source of Salivary Stem/Progenitor Cells for Regenerative Applications.
Methods in molecular biology (Clifton, N.J.) (2023-12-22)
Caitlynn M L Barrows, Danielle Wu, Simon Young, Mary C Farach-Carson
ABSTRACT
Resident stem/progenitor cells within the secretory salivary glands offer a potential therapeutic resource for use in the regeneration of salivary glands needed to restore saliva production in patients with chronic xerostomia, or dry mouth. Methods were developed previously to isolate human stem/progenitor cells (hS/PCs) from major salivary glands (parotid/submandibular). Abundant minor salivary glands located in readily accessible locations in the oral cavity and lip could provide an additional valuable therapeutic resource. An advantage of this cell resource is that these minor glands about the size of grape seeds can be harvested from healthy donors using minimally invasive surgical procedures. The disadvantage of using minor glands is that they contain many fewer cells than do major glands, and thus harvested cells need to be expanded in the lab to create a therapeutic resource. While earlier work has described isolation of proliferative cell populations from minor salivary glands that could be used in regenerative medicine, most of these expanded cells possess properties of mesenchymal cells rather than the epithelial population that secretes salivary products.Here, we describe in detail our recently established methods to isolate and expand hS/PCs isolated from human labial minor salivary glands. Expanded hS/PC populations are epithelial assessed by their expression of epithelial progenitor markers K5 and K14. Like expandable cell populations previously isolated from the major salivary glands, these cells also express nuclear p63, consistent with their ability to be expanded after explant culture. When hS/PCs with these properties are encapsulated into a customized 3D biomimetic hyaluronic acid-based hydrogel, they will assemble into microstructures that retain some progenitor markers while also beginning to differentiate. The increased expression of secreted mucin MUC-7 was used to demonstrate differentiation and secretory potential in assembled hS/PC microstructures. Compared to hS/PCs from major glands, those from minor salivary glands tend to be more heterogeneous in early passage; thus, use of K5/K14/p63 as an early quality assessment tool is highly recommended. Additionally, hS/PCs from minor glands are sensitive to stress and if mishandled will demonstrate a stress response that leads to their transitioning to a flat, squamous cell-like appearance that is of limited utility in regenerative medicine applications. We conclude that properly handled hS/PCs from minor salivary glands represent a powerful new source of therapeutic cells for applications including treating patients with chronic xerostomia.
Sigma-Aldrich
Anti-Cytokeratin 14 Antibody, clone LL002, clone LL002, Chemicon®, from mouse