Merck
  • Disruption of Cav1.2-mediated signaling is a pathway for ketamine-induced pathology.
Disruption of Cav1.2-mediated signaling is a pathway for ketamine-induced pathology.
Nature communications (2020-08-30)
Huan Chen, David H Vandorpe, Xiang Xie, Seth L Alper, Mark L Zeidel, Weiqun Yu
ABSTRACT
The general anesthetic ketamine has been repurposed by physicians as an anti-depressant and by the public as a recreational drug. However, ketamine use can cause extensive pathological changes, including ketamine cystitis. The mechanisms of ketamine's anti-depressant and adverse effects remain poorly understood. Here we present evidence that ketamine is an effective L-type Ca2+ channel (Cav1.2) antagonist that directly inhibits calcium influx and smooth muscle contractility, leading to voiding dysfunction. Ketamine prevents Cav1.2-mediated induction of immediate early genes and transcription factors, and inactivation of Cav1.2 in smooth muscle mimics the ketamine cystitis phenotype. Our results demonstrate that ketamine inhibition of Cav1.2 signaling is an important pathway mediating ketamine cystitis. In contrast, Cav1.2 agonist Bay k8644 abrogates ketamine-induced smooth muscle dysfunction. Indeed, Cav1.2 activation by Bay k8644 decreases voiding frequency while increasing void volume, indicating Cav1.2 agonists might be effective drugs for treatment of bladder dysfunction.
Sigma-Aldrich
Anti-Calcium Channel Antibody, Voltage Gated α 1C, Chemicon®, from rabbit