Merck
  • Rapamycin-induced G1 cell cycle arrest employs both TGF-β and Rb pathways.
Rapamycin-induced G1 cell cycle arrest employs both TGF-β and Rb pathways.
Cancer letters (2015-02-11)
Amrita Chatterjee, Suman Mukhopadhyay, Kaity Tung, Deven Patel, David A Foster
ABSTRACT
The mammalian target of rapamycin complex 1 (mTORC1) is a critical regulator of G1 cell cycle progression. Two key substrates of mTORC1 are ribosomal subunit S6 kinase (S6K) and eukaryotic initiation factor 4E (eIF4E) binding protein-1 (4E-BP1). We reported previously that simultaneous knockdown of S6K and eIF4E causes a transforming growth factor-β (TGF-β)-dependent G1 cell cycle arrest in MDA-MB-231 human breast cancer cells. Rapamycin inhibits the phosphorylation of S6K at nano-molar concentrations in MDA-MB-231 cells; however, micro-molar concentrations of rapamycin are required to inhibit phosphorylation of 4E-BP1 - the phosphorylation of which liberates eIF4E to initiate translation. Micro-molar doses of rapamycin are required for complete G1 cell cycle arrest - indicating that 4E-BP1 is a critical target of mTOR for promoting cell cycle progression. Data are provided demonstrating that G1 cell cycle arrest induced by rapamycin is due to up-regulation of TGF-β signaling and down-regulation of Rb phosphorylation via phosphorylation of the mTORC1 substrates S6K and 4E-BP1 respectively. These findings enhance the current understanding of the cytostatic effects of mTORC1 suppression with therapeutic implications.
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-phospho-Smad2 (Ser465/467) Antibody, clone A5S, rabbit monoclonal, culture supernatant, clone A5S, Upstate®