
Tristearin

Synonym(s):
Glyceryl tristearate, 1,2,3-Trioctadecanoylglycerol, Glycerol tristearate, Tristearin
Linear Formula:
[CH3(CH2)16COOCH2]2CHOCO(CH2)16CH3
CAS Number:
Molecular Weight:
891.48
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing