
Rabeprazole for system suitability

Synonym(s):
Rabeprazole sodium, 2-([4-(3-Methoxypropoxy)-3-methylpyridin-2-yl]methylsulfinyl)-1H-benzo[d]imidazole, Pariprazole
Empirical Formula (Hill Notation):
C18H20N3O3S · Na
CAS Number:
Molecular Weight:
381.42
Products
Formula Weight
Gene Alias
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing