
Pilocarpine hydrochloride

Synonym(s):
(3S,4R)-4,5-Dihydro-3-ethyl-4-(1-methyl-1H-imidazol-5-ylmethyl)-2(3H)-furanone hydrochloride, Pilocarpine hydrochloride
Empirical Formula (Hill Notation):
C11H16N2O2 · HCl
CAS Number:
Molecular Weight:
244.72
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Relevant Disease
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing