
Phosmet oxon

Synonym(s):
S-[(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)methyl] O,O-dimethyl phosphorothioate
Empirical Formula (Hill Notation):
C11H12NO5PS
CAS Number:
Molecular Weight:
301.26
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing