
PCB No 138

Synonym(s):
2,2′,3,4,4′,5′-Hexachlorobiphenyl
Empirical Formula (Hill Notation):
C12H4Cl6
CAS Number:
Molecular Weight:
360.88
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing