![Orbifloxacin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/587/485/fa2be7d4-7f1d-419a-82a0-1f1aa75805f9/640/fa2be7d4-7f1d-419a-82a0-1f1aa75805f9.png)
Orbifloxacin
![Orbifloxacin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/587/485/fa2be7d4-7f1d-419a-82a0-1f1aa75805f9/640/fa2be7d4-7f1d-419a-82a0-1f1aa75805f9.png)
Synonym(s):
1-Cyclopropyl-5,6,8-trifluoro-1,4-dihydro-7-(cis-3,5-dimethyl-1-piperazinyl)-4-oxo-quinoline-3-carboxylic acid, Orbax
Empirical Formula (Hill Notation):
C19H20F3N3O3
CAS Number:
Molecular Weight:
395.38
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3