
Olanzapine Related Compound B

Synonym(s):
5,10-Dihydro-2-methyl-4H-thieno[2,3-b][1,5]benzodiazepin-4-one, 2-Methyl-4,9-dihydro-3-thia-4,9-diazabenzo[f]azulene-10-one, Olanzapine lactam
Empirical Formula (Hill Notation):
C12H10N2OS
CAS Number:
Molecular Weight:
230.29
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2